Assay Details
| Gene Symbol: CALML4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685149.3 | XP_005685206.1 | 4 - 5 | 1988 | 76 | |
| XM_005685150.3 | XP_005685207.2 | 3 - 4 | 267 | 76 | ||
| XM_013966779.2 | XP_013822233.1 | 3 - 4 | 711 | 76 | ||
| GenBank mRNA | JO437376.2 | - | 4 - 5 | 786 | 76 | |
More Information
Set Membership: |
|

CALML4
Genomic Map