Assay Details
| Gene Symbol: HAUS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685258.3 | XP_005685315.2 | 3 - 4 | 592 | 61 | |
| XM_005685259.3 | XP_005685316.2 | 3 - 4 | 382 | 61 | ||
| XM_018054576.1 | XP_017910065.1 | 3 - 4 | 545 | 61 | ||
| XM_018054578.1 | XP_017910067.1 | 3 - 4 | 541 | 61 | ||
| XM_018054579.1 | XP_017910068.1 | 3 - 4 | 410 | 61 | ||
| GenBank mRNA | JO499480.2 | - | 3 - 4 | 471 | 61 | |
More Information
Set Membership: |
|

HAUS4
Genomic Map