Assay Details
| Gene Symbol: PROCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688489.3 | XP_005688546.1 | 2 - 3 | 211 | 62 | |
| XM_013968973.2 | XP_013824427.1 | 2 - 3 | 239 | 62 | ||
| XM_013968974.2 | XP_013824428.1 | 1 - 2 | 258 | 62 | ||
| XM_018057757.1 | XP_017913246.1 | 1 - 2 | 258 | 62 | ||
| GenBank mRNA | JO350568.2 | - | 2 - 3 | 162 | 62 | |
More Information
Set Membership: |
|

PROCR
Genomic Map