Assay Details
| Gene Symbol: LOC102173760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057769.1 | XP_017913258.1 | 18 - 19 | 2304 | 64 | |
| XM_018057770.1 | XP_017913259.1 | 18 - 19 | 2304 | 64 | ||
| XM_018057771.1 | XP_017913260.1 | 18 - 19 | 2304 | 64 | ||
| XM_018057772.1 | XP_017913261.1 | 18 - 19 | 2304 | 64 | ||
More Information
Set Membership: |
|

LOC102173760
Genomic Map