Assay Details
| Gene Symbol: LOC102172279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058445.1 | XP_017913934.1 | 15 - 16 | 1464 | 93 | |
| XM_018058446.1 | XP_017913935.1 | 15 - 16 | 1449 | 93 | ||
| XM_018058447.1 | XP_017913936.1 | 14 - 15 | 1380 | 93 | ||
More Information
Set Membership: |
|

LOC102172279
Genomic Map