Assay Details
| Gene Symbol: LOC102172279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058445.1 | XP_017913934.1 | 10 - 11 | 971 | 147 | |
| XM_018058446.1 | XP_017913935.1 | 10 - 11 | 956 | 147 | ||
| XM_018058447.1 | XP_017913936.1 | 9 - 10 | 887 | 147 | ||
More Information
Set Membership: |
|

LOC102172279
Genomic Map