Assay Details
| Gene Symbol: PABPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058311.1 | XP_017913800.1 | 10 - 11 | 1973 | 63 | |
| XM_018058312.1 | XP_017913801.1 | 8 - 9 | 1740 | 63 | ||
| GenBank mRNA | JO581303.1 | - | 10 - 11 | 2050 | 63 | |
More Information
Set Membership: |
|

PABPC1
Genomic Map