Assay Details
| Gene Symbol: TSEN15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690969.3 | XP_005691026.3 | 3 - 4 | 418 | 65 | |
| GenBank mRNA | JO603770.2 | - | 3 - 4 | 422 | 65 | |
More Information
Set Membership: |
|

TSEN15
Genomic Map