Assay Details
| Gene Symbol: NUFIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693278.3 | XP_005693335.1 | 2 - 3 | 2120 | 77 | |
| GenBank mRNA | JO329894.2 | - | 2 - 3 | 1970 | 77 | |
| JO648702.2 | - | 1 - 2 | 380 | 77 | ||
More Information
Set Membership: |
|

NUFIP2
Genomic Map