Assay Details
| Gene Symbol: GEMIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693304.3 | XP_005693361.2 | 1 - 2 | 239 | 121 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005693304.3 transcripts.
Set Membership: |
|

GEMIN4
Genomic Map