Assay Details
| Gene Symbol: C19H17orf49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064536.1 | XP_017920025.1 | 2 - 3 | 352 | 68 | |
| XM_018064537.1 | XP_017920026.1 | 2 - 3 | 352 | 68 | ||
| GenBank mRNA | JO351749.2 | - | 4 - 5 | 656 | 68 | |
| JO611484.2 | - | 2 - 3 | 230 | 68 | ||
| KA318145.1 | - | 2 - 3 | 295 | 68 | ||
More Information
Set Membership: |
|

C19H17orf49
Genomic Map