Assay Details
| Gene Symbol: G6PC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693916.3 | XP_005693973.1 | 1 - 2 | 471 | 62 | |
| XM_018065207.1 | XP_017920696.1 | 1 - 2 | 471 | 62 | ||
| GenBank mRNA | JO621445.2 | - | 1 - 2 | 449 | 62 | |
| JO621447.2 | - | 1 - 2 | 449 | 62 | ||
More Information
Set Membership: |
|

G6PC3
Genomic Map