Assay Details
| Gene Symbol: LOC100861338 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697733.3 | XP_005697790.1 | 2 - 3 | 439 | 59 | |
| GenBank mRNA | JO582241.1 | - | 1 - 2 | 598 | 59 | |
| KM576768.1 | - | 2 - 3 | 332 | 59 | ||
| KM576770.1 | - | 2 - 3 | 332 | 59 | ||
More Information
Set Membership: |
|

LOC100861338
Genomic Map