Assay Details
| Gene Symbol: NACAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047040.1 | XP_017902529.1 | 2 - 3 | 6597 | 55 | |
| XM_018047041.1 | XP_017902530.1 | 2 - 3 | 6597 | 55 | ||
| XM_018047042.1 | XP_017902531.1 | 3 - 4 | 5746 | 55 | ||
| GenBank mRNA | JT665620.1 | - | 1 - 2 | 48 | 55 | |
More Information
Set Membership: |
|

NACAD
Genomic Map