Assay Details
| Gene Symbol: CBARP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050459.1 | XP_017905948.1 | 3 - 4 | 2299 | 54 | |
| XM_018050460.1 | XP_017905949.1 | 2 - 3 | 856 | 54 | ||
| XM_018050461.1 | XP_017905950.1 | 3 - 4 | 2299 | 54 | ||
| GenBank mRNA | JO638876.2 | - | 7 - 8 | 3026 | 54 | |
| KA335381.1 | - | 5 - 6 | 625 | 54 | ||
More Information
Set Membership: |
|

CBARP
Genomic Map