Assay Details
| Gene Symbol: LOC102181119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013965615.2 | XP_013821069.2 | 2 - 3 | 3237 | 76 | |
| XM_018050258.1 | XP_017905747.1 | 2 - 3 | 2579 | 76 | ||
| XM_018050259.1 | XP_017905748.1 | 2 - 3 | 2492 | 76 | ||
| XM_018050260.1 | XP_017905749.1 | 2 - 3 | 3237 | 76 | ||
| XM_018050261.1 | XP_017905750.1 | 2 - 3 | 2571 | 76 | ||
| XM_018050262.1 | XP_017905751.1 | 2 - 3 | 2542 | 76 | ||
More Information
Set Membership: |
|

LOC102181119
Genomic Map