Assay Details
| Gene Symbol: TMEM62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013967012.2 | XP_013822466.1 | 1 - 2 | 518 | 68 | |
| XM_018054381.1 | XP_017909870.1 | 1 - 2 | 519 | 68 | ||
| GenBank mRNA | JO427696.1 | - | 13 - 14 | 2313 | 68 | |
| KA335476.1 | - | 1 - 2 | 481 | 68 | ||
More Information
Set Membership: |
|

TMEM62
Genomic Map