Assay Details
| Gene Symbol: ANGPTL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055801.1 | XP_017911290.1 | 3 - 4 | 1736 | 60 | |
| GenBank mRNA | JO497102.2 | - | 2 - 3 | 1968 | 60 | |
| KA339063.1 | - | 3 - 4 | 1479 | 60 | ||
More Information
Set Membership: |
|

ANGPTL2
Genomic Map