Assay Details
| Gene Symbol: TASOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013968582.2 | XP_013824036.2 | 13 - 14 | 1833 | 140 | |
| XM_013968583.2 | XP_013824037.2 | 13 - 14 | 1851 | 140 | ||
| XM_018057295.1 | XP_017912784.1 | 11 - 12 | 1543 | 140 | ||
| XM_018057296.1 | XP_017912785.1 | 13 - 14 | 1851 | 140 | ||
| XM_018057297.1 | XP_017912786.1 | 6 - 7 | 565 | 140 | ||
| XM_018057298.1 | XP_017912787.1 | 13 - 14 | 1833 | 140 | ||
More Information
Set Membership: |
|

TASOR2
Genomic Map