Assay Details
| Gene Symbol: TASOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013968582.2 | XP_013824036.2 | 16 - 17 | 2693 | 98 | |
| XM_018057295.1 | XP_017912784.1 | 14 - 15 | 2403 | 98 | ||
| XM_018057296.1 | XP_017912785.1 | 16 - 17 | 2711 | 98 | ||
| XM_018057297.1 | XP_017912786.1 | 9 - 10 | 1425 | 98 | ||
More Information
Set Membership: |
|

TASOR2
Genomic Map