Assay Details
| Gene Symbol: TASOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013968582.2 | XP_013824036.2 | 19 - 20 | 4220 | 103 | |
| XM_013968583.2 | XP_013824037.2 | 18 - 19 | 3995 | 103 | ||
| XM_018057295.1 | XP_017912784.1 | 18 - 19 | 4119 | 103 | ||
| XM_018057296.1 | XP_017912785.1 | 19 - 20 | 4238 | 103 | ||
| XM_018057297.1 | XP_017912786.1 | 13 - 14 | 3141 | 103 | ||
| XM_018057298.1 | XP_017912787.1 | 18 - 19 | 3977 | 103 | ||
More Information
Set Membership: |
|

TASOR2
Genomic Map