Assay Details
| Gene Symbol: TASOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013968582.2 | XP_013824036.2 | 21 - 22 | 8097 | 88 | |
| XM_013968583.2 | XP_013824037.2 | 20 - 21 | 7872 | 88 | ||
| XM_018057295.1 | XP_017912784.1 | 20 - 21 | 7996 | 88 | ||
| XM_018057296.1 | XP_017912785.1 | 21 - 22 | 8115 | 88 | ||
| XM_018057297.1 | XP_017912786.1 | 15 - 16 | 7018 | 88 | ||
| XM_018057298.1 | XP_017912787.1 | 20 - 21 | 7854 | 88 | ||
| GenBank mRNA | JT681980.1 | - | 7 - 8 | 1324 | 88 | |
More Information
Set Membership: |
|

TASOR2
Genomic Map