Assay Details
| Gene Symbol: TASOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013968582.2 | XP_013824036.2 | 5 - 6 | 1075 | 85 | |
| XM_013968583.2 | XP_013824037.2 | 5 - 6 | 1075 | 85 | ||
| XM_018057295.1 | XP_017912784.1 | 3 - 4 | 767 | 85 | ||
| XM_018057296.1 | XP_017912785.1 | 5 - 6 | 1075 | 85 | ||
| XM_018057298.1 | XP_017912787.1 | 5 - 6 | 1075 | 85 | ||
More Information
Set Membership: |
|

TASOR2
Genomic Map