Assay Details
| Gene Symbol: LOC102174762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058179.1 | XP_017913668.1 | 2 - 3 | 395 | 93 | |
| GenBank mRNA | JO612734.2 | - | 6 - 7 | 1007 | 93 | |
| JO646076.2 | - | 2 - 3 | 151 | 93 | ||
More Information
Set Membership: |
|

LOC102174762
Genomic Map