Assay Details
| Gene Symbol: LOC102168948 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063538.1 | XP_017919027.1 | 31 - 32 | 4649 | 103 | |
| XM_018063553.1 | XP_017919042.1 | 31 - 32 | 4649 | 103 | ||
More Information
Set Membership: |
|

LOC102168948
Genomic Map