Assay Details
| Gene Symbol: GPR153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060443.1 | XP_017915932.1 | 4 - 5 | 1284 | 74 | |
| XM_018060444.1 | XP_017915933.1 | 4 - 5 | 1470 | 74 | ||
| GenBank mRNA | JO645663.2 | - | 3 - 4 | 1562 | 74 | |
More Information
Set Membership: |
|

GPR153
Genomic Map