Assay Details
| Gene Symbol: WRAP73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060462.1 | XP_017915951.1 | 4 - 5 | 558 | 59 | |
| XM_018060463.1 | XP_017915952.1 | 3 - 4 | 415 | 59 | ||
| XM_018060464.1 | XP_017915953.1 | 4 - 5 | 561 | 59 | ||
| GenBank mRNA | JO587006.2 | - | 3 - 4 | 452 | 59 | |
More Information
Set Membership: |
|

WRAP73
Genomic Map