Assay Details
| Gene Symbol: MAP2K4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064671.1 | XP_017920160.1 | 8 - 9 | 913 | 69 | |
| XM_018064672.1 | XP_017920161.1 | 7 - 8 | 851 | 69 | ||
More Information
Set Membership: |
|

MAP2K4
Genomic Map