Assay Details
| Gene Symbol: RAB26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040984.1 | XP_017896473.1 | 4 - 5 | 565 | 66 | |
| XM_018040985.1 | XP_017896474.1 | 4 - 5 | 565 | 66 | ||
| GenBank mRNA | JO454750.2 | - | 1 - 2 | 78 | 66 | |
| JT670585.1 | - | 3 - 4 | 221 | 66 | ||
More Information
Set Membership: |
|

RAB26
Genomic Map