Assay Details
| Gene Symbol: LOC108637091 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054726.1 | XP_017910215.1 | 2054 | 94 | ||
| GenBank mRNA | JO580388.1 | - | 665 | 94 | ||
| JO634691.2 | - | 1759 | 94 | |||
| Gene Symbol: LOC102173345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054721.1 | XP_017910210.1 | 1850 | 94 | ||
More Information
Set Membership: |
|

LOC108637091
Genomic Map