Assay Details
| Gene Symbol: LOC108637091 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO580388.1 | - | 1264 | 62 | ||
| JO634691.2 | - | 1160 | 62 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC108637091| Gene Symbol: LOC108637091 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO580388.1 | - | 1264 | 62 | ||
| JO634691.2 | - | 1160 | 62 | |||
Set Membership: |
|