Assay Details
| Gene Symbol: LOC108637091 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054726.1 | XP_017910215.1 | 287 | 114 | ||
| GenBank mRNA | JO634691.2 | - | 3526 | 114 | ||
| Gene Symbol: LOC102173345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054721.1 | XP_017910210.1 | 98 | 112 | ||
| Gene Symbol: LOC108635191 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045451.1 | XP_017900940.1 | 941 | 114 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018054721.1 transcripts.
Set Membership: |
|

LOC108637091
Genomic Map