Assay Details
| Gene Symbol: CD247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013975200.2 | XP_013830654.2 | 6 - 7 | 559 | 59 | |
| XM_018046444.1 | XP_017901933.1 | 6 - 7 | 559 | 59 | ||
| GenBank mRNA | JO581444.1 | - | 6 - 7 | 722 | 59 | |
More Information
Set Membership: |
|

CD247
Genomic Map