Assay Details
| Gene Symbol: LSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042933.1 | XP_017898422.1 | 7 - 8 | 1006 | 84 | |
| XM_018042934.1 | XP_017898423.1 | 7 - 8 | 1006 | 84 | ||
| XM_018042935.1 | XP_017898424.1 | 7 - 8 | 1006 | 84 | ||
| XM_018042937.1 | XP_017898426.1 | 7 - 8 | 1006 | 84 | ||
| XM_018042938.1 | XP_017898427.1 | 7 - 8 | 1006 | 84 | ||
| XM_018042939.1 | XP_017898428.1 | 7 - 8 | 1006 | 84 | ||
| XM_018042940.1 | XP_017898429.1 | 7 - 8 | 1006 | 84 | ||
| XM_018042941.1 | XP_017898430.1 | 7 - 8 | 1269 | 84 | ||
| XM_018042942.1 | XP_017898431.1 | 7 - 8 | 999 | 84 | ||
| XM_018042943.1 | XP_017898432.1 | 7 - 8 | 842 | 84 | ||
| XM_018042944.1 | XP_017898433.1 | 7 - 8 | 842 | 84 | ||
| XM_018042945.1 | XP_017898434.1 | 7 - 8 | 1333 | 84 | ||
| XM_018042946.1 | XP_017898435.1 | 7 - 8 | 784 | 84 | ||
| GenBank mRNA | JO433952.2 | - | 4 - 5 | 821 | 84 | |
More Information
Set Membership: |
|

LSP1
Genomic Map