Assay Details
| Gene Symbol: PCP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675627.3 | XP_005675684.1 | 2 - 3 | 162 | 83 | |
| GenBank mRNA | KA350601.1 | - | 2 - 3 | 129 | 83 | |
More Information
Set Membership: |
|

PCP4
Genomic Map