Assay Details
| Gene Symbol: ITPRID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065361.1 | XP_017920850.1 | 11 - 12 | 3134 | 82 | |
| XM_018065369.1 | XP_017920858.1 | 11 - 12 | 3021 | 82 | ||
| XM_018065378.1 | XP_017920867.1 | 11 - 12 | 3134 | 82 | ||
| GenBank mRNA | JO401797.2 | - | 7 - 8 | 1227 | 82 | |
| KA324907.1 | - | 1821 | 82 | |||
More Information
Set Membership: |
|

ITPRID2
Genomic Map