Assay Details
| Gene Symbol: LOC102169350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678019.2 | XP_005678076.1 | 9 - 10 | 1554 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102169350| Gene Symbol: LOC102169350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678019.2 | XP_005678076.1 | 9 - 10 | 1554 | 100 | |
Set Membership: |
|