Assay Details
| Gene Symbol: LRRC39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678047.3 | XP_005678104.2 | 6 - 7 | 877 | 86 | |
| XM_018045818.1 | XP_017901307.1 | 6 - 7 | 778 | 86 | ||
| XM_018045819.1 | XP_017901308.1 | 6 - 7 | 881 | 86 | ||
| GenBank mRNA | JO368474.2 | - | 3 - 4 | 494 | 86 | |
More Information
Set Membership: |
|

LRRC39
Genomic Map