Assay Details
| Gene Symbol: ABCF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679657.3 | XP_005679714.1 | 13 - 14 | 1854 | 57 | |
| XM_018046710.1 | XP_017902199.1 | 13 - 14 | 1793 | 57 | ||
More Information
Set Membership: |
|

ABCF2
Genomic Map