Assay Details
| Gene Symbol: LOC102168522 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049530.1 | XP_017905019.1 | 2 - 3 | 890 | 76 | |
| XM_018049531.1 | XP_017905020.1 | 2 - 3 | 903 | 76 | ||
| XM_018049532.1 | XP_017905021.1 | 3 - 4 | 981 | 76 | ||
| GenBank mRNA | JO561351.2 | - | 6 - 7 | 976 | 76 | |
More Information
Set Membership: |
|

LOC102168522
Genomic Map