Assay Details
| Gene Symbol: EPGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681753.2 | XP_005681810.1 | 3 - 4 | 318 | 78 | |
| GenBank mRNA | KA314941.1 | - | 2 - 3 | 201 | 78 | |
More Information
Set Membership: |
|

EPGN
Genomic Map