Assay Details
| Gene Symbol: MRPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681806.3 | XP_005681863.2 | 1 - 2 | 725 | 55 | |
| XM_018049624.1 | XP_017905113.1 | 1 - 2 | 725 | 55 | ||
More Information
Set Membership: |
|

MRPL1
Genomic Map