Assay Details
Gene Symbol: UBLCP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005683237.3 | XP_005683294.1 | 8 - 9 | 1762 | 78 | |
GenBank mRNA | JO576576.1 | - | 3 - 4 | 1364 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
