Assay Details
| Gene Symbol: CCNH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683411.3 | XP_005683468.1 | 1 - 2 | 285 | 69 | |
| XM_005683413.3 | XP_005683470.1 | 1 - 2 | 282 | 69 | ||
| XM_005683414.3 | XP_005683471.1 | 1 - 2 | 284 | 69 | ||
| XM_005683415.3 | XP_005683472.1 | 1 - 2 | 282 | 69 | ||
More Information
Set Membership: |
|

CCNH
Genomic Map