Assay Details
| Gene Symbol: DCAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686037.3 | XP_005686094.2 | 13 - 14 | 1436 | 63 | |
| XM_005686038.3 | XP_005686095.2 | 13 - 14 | 1433 | 63 | ||
| GenBank mRNA | JO430991.2 | - | 13 - 14 | 1454 | 63 | |
More Information
Set Membership: |
|

DCAF4
Genomic Map