Assay Details
| Gene Symbol: NDUFA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687038.2 | XP_005687095.1 | 3 - 4 | 516 | 101 | |
| GenBank mRNA | JO642951.2 | - | 3 - 4 | 500 | 101 | |
More Information
Set Membership: |
|

NDUFA8
Genomic Map