Assay Details
| Gene Symbol: SLA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688519.3 | XP_005688576.2 | 7 - 8 | 1098 | 63 | |
| XM_013968921.2 | XP_013824375.2 | 7 - 8 | 1110 | 63 | ||
| GenBank mRNA | JO318731.2 | - | 4 - 5 | 2009 | 63 | |
| JO582119.1 | - | 4 - 5 | 2123 | 63 | ||
More Information
Set Membership: |
|

SLA2
Genomic Map