Assay Details
| Gene Symbol: DMAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692438.3 | XP_005692495.2 | 4 - 5 | 463 | 78 | |
| GenBank mRNA | KA325771.1 | - | 2 - 3 | 1352 | 78 | |
More Information
Set Membership: |
|

DMAC2
Genomic Map