Assay Details
| Gene Symbol: CCL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693201.3 | XP_005693258.1 | 1 - 2 | 254 | 99 | |
| GenBank mRNA | JO333602.2 | - | 2 - 3 | 587 | 99 | |
| JT762860.1 | - | 534 | 99 | |||
More Information
Set Membership: |
|

CCL5
Genomic Map