Assay Details
| Gene Symbol: SLC1A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694788.3 | XP_005694845.2 | 7 - 8 | 1319 | 62 | |
| XM_018065700.1 | XP_017921189.1 | 5 - 6 | 866 | 62 | ||
More Information
Set Membership: |
|

SLC1A3
Genomic Map